siRNA Details
Gene Symbol: Fmnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105846.1 | NP_001099316.1 | 3 | 450 | ||
XM_006247498.2 | XP_006247560.1 | 3 | 661 | |||
XM_006247499.2 | XP_006247561.1 | 3 | 662 | |||
XM_006247500.2 | XP_006247562.1 | 3 | 661 | |||
XM_006247501.2 | XP_006247563.1 | 3 | 661 | |||
XM_008768266.1 | XP_008766488.1 | 3 | 660 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
