siRNA Details
Gene Symbol: Smndc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025400.3 | NP_001020571.1 | 2, 3 | 155 | ||
NM_001035234.2 | NP_001030311.1 | 2, 3 | 317 | |||
XM_006231608.2 | XP_006231670.1 | 2, 3 | 594 | |||
GenBank mRNA | BC097986.1 | - | 2, 3 | 155 | 7444662 | |
BC098697.1 | - | 2, 3 | 279 | 7386640 | ||
EV778425.1 | - | 2, 3 | 317 | |||
FQ220088.1 | - | 1, 2 | 98 |
Gene Symbol: LOC100363406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768448.1 | XP_008766670.1 | 1, 2 | 280 |
