siRNA Details
Gene Symbol: Abca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768405.1 | XP_008766627.1 | 36, 37 | 5726 |
Gene Symbol: LOC102553158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768451.1 | XP_008766673.1 | 335 |
