siRNA Details
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025111.1 | NP_001020282.1 | 2 | 390 | ||
XM_008768332.1 | XP_008766554.1 | 3 | 1246 | |||
XM_008768333.1 | XP_008766555.1 | 4 | 775 | |||
XM_008768334.1 | XP_008766556.1 | 3 | 665 | |||
XM_008768335.1 | XP_008766557.1 | 2 | 728 | |||
XM_008768336.1 | XP_008766558.1 | 2 | 494 | |||
XM_008768337.1 | XP_008766559.1 | 3 | 1246 | |||
XM_008768338.1 | XP_008766560.1 | 3 | 1244 | |||
XM_008768339.1 | XP_008766561.1 | 3 | 1240 | |||
XM_008768340.1 | XP_008766562.1 | 3 | 1243 | |||
XM_008768341.1 | XP_008766563.1 | 3 | 1238 | |||
XM_008768342.1 | XP_008766564.1 | 3 | 1235 | |||
GenBank mRNA | BC093593.1 | - | 2 | 390 | 7314621 | |
GU057984.1 | - | 2 | 358 | |||
GU062903.1 | - | 2 | 355 | |||
HQ263412.1 | - | 2 | 358 |
