siRNA Details
Gene Symbol: Fbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247750.2 | XP_006247812.1 | 11 | 1523 | ||
XM_006247759.2 | XP_006247821.1 | 11 | 947 | |||
XM_006247760.2 | XP_006247822.1 | 11 | 1523 | |||
XM_006247761.2 | XP_006247823.1 | 11 | 1523 | |||
XM_008768413.1 | XP_008766635.1 | 11 | 1520 | |||
XM_008768414.1 | XP_008766636.1 | 11 | 1523 | |||
XM_008768415.1 | XP_008766637.1 | 11 | 1524 | |||
XM_008768416.1 | XP_008766638.1 | 12 | 1598 | |||
XM_008768417.1 | XP_008766639.1 | 12 | 1026 | |||
XM_008768418.1 | XP_008766640.1 | 924 | ||||
XM_008768419.1 | XP_008766641.1 | 12 | 1595 | |||
XM_008768420.1 | XP_008766642.1 | 11 | 1524 | |||
XM_008768421.1 | XP_008766643.1 | 4 | 560 | |||
GenBank mRNA | FQ210392.1 | - | 11 | 885 |
