siRNA Details
Gene Symbol: Actg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127449.1 | NP_001120921.1 | 4 | 696 | ||
GenBank mRNA | CF110974.1 | - | 1 | 270 | ||
CK364554.1 | - | 3 | 612 | |||
FQ216158.1 | - | 5 | 737 | |||
FQ227152.1 | - | 1 | 167 | |||
FQ230767.1 | - | 1 | 167 | |||
FQ231596.1 | - | 1 | 167 | |||
FQ231807.1 | - | 1 | 252 | |||
X52815.1 | - | 3 | 645 |
Gene Symbol: LOC100361457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729196.3 | XP_002729242.2 | 1 | 693 |
Gene Symbol: Potef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728532.4 | XP_002728578.1 | 1 | 692 |

More Information
Other Species Targeted

Gene Ontology Categories:
