siRNA Details
Gene Symbol: Top3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105861.1 | NP_001099331.1 | 5 | 504 | ||
XM_006248671.2 | XP_006248733.1 | 4 | 638 | |||
XM_006248673.2 | XP_006248735.1 | 4 | 450 | |||
GenBank mRNA | FQ211004.1 | - | 4 | 450 | ||
FQ212059.1 | - | 4 | 459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
