siRNA Details
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013863.2 | NP_001013885.2 | 5 | 1014 | ||
XM_008768859.1 | XP_008767081.1 | 6 | 1047 | |||
XM_008768860.1 | XP_008767082.1 | 7 | 1463 | |||
XM_008768861.1 | XP_008767083.1 | 7 | 1212 | |||
GenBank mRNA | BC086565.1 | - | 5 | 1164 | 7113479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
