siRNA Details
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105864.1 | NP_001099334.1 | 5 | 1106 | ||
XM_006248688.2 | XP_006248750.1 | 5 | 1094 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153790, AK170331, AK142378, AB024432, AK143208, and others.
Gene Ontology Categories:
