siRNA Details
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011895.1 | NP_001011895.1 | 2 | 690 | ||
XM_006248604.1 | XP_006248666.1 | 2 | 902 | |||
XM_006248605.1 | XP_006248667.1 | 2 | 661 | |||
GenBank mRNA | BC083619.1 | - | 2 | 690 | 7133938 |
