siRNA Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013864.1 | NP_001013886.1 | 5, 6 | 546 | ||
XM_006248505.2 | XP_006248567.1 | 8, 9 | 983 | |||
GenBank mRNA | BC083627.1 | - | 5, 6 | 546 | 7134282 | |
BC083890.1 | - | 4, 5 | 484 | 7132481 |
