siRNA Details
Gene Symbol: Fam43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039002.1 | NP_001034091.1 | 1 | 1315 | ||
GenBank mRNA | BC099231.1 | - | 1 | 1315 | 7442928 |
Gene Symbol: LOC100909474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752202.3 | XP_003752250.1 | 1 | 1140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
