siRNA Details
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105874.2 | NP_001099344.2 | 28 | 5252 | ||
XM_006248433.2 | XP_006248495.1 | 28 | 5560 | |||
XM_006248434.2 | XP_006248496.1 | 28 | 5342 | |||
XM_008768711.1 | XP_008766933.1 | 28 | 5561 | |||
XM_008768712.1 | XP_008766934.1 | 12 | 1972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
