siRNA Details
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271250.1 | NP_001258179.1 | 11 | 1299 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001114611, NM_001114613, NM_001114612, NM_172440, AK161955, and others.
Gene Ontology Categories:
