siRNA Details
Gene Symbol: RGD1566314 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248319.2 | XP_006248381.1 | 7 | 1116 | ||
XM_008768763.1 | XP_008766985.1 | 7 | 1116 | |||
XM_008768764.1 | XP_008766986.1 | 7 | 1116 |
