siRNA Details
Gene Symbol: Naa50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105881.1 | NP_001099351.1 | 557 | |||
XM_006248326.2 | XP_006248388.1 | 3, 4 | 563 | |||
GenBank mRNA | BC169000.1 | - | 3, 4 | 553 | 8362219 | |
FQ216855.1 | - | 3, 4 | 473 |
Gene Symbol: LOC102553099 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248358.2 | XP_006248420.1 | 3, 4 | 316 |

More Information
Other Species Targeted

Gene Ontology Categories:
