siRNA Details
Gene Symbol: Sidt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100653.1 | NP_001094123.1 | 5 | 642 | ||
XM_006248328.2 | XP_006248390.1 | 5 | 1521 | |||
XM_008768718.1 | XP_008766940.1 | 5 | 1520 | |||
GenBank mRNA | AY562215.1 | - | 5 | 642 |
