siRNA Details
Gene Symbol: Spice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008285.1 | NP_001008286.1 | 14, 15 | 2193 | ||
XM_006248331.2 | XP_006248393.1 | 15, 16 | 2407 | |||
XM_006248332.2 | XP_006248394.1 | 14, 15 | 2361 | |||
XM_006248334.1 | XP_006248396.1 | 12, 13 | 2090 | |||
XM_008768719.1 | XP_008766941.1 | 14, 15 | 2291 | |||
XM_008768721.1 | XP_008766943.1 | 12, 13 | 2622 | |||
GenBank mRNA | BC085954.1 | - | 14, 15 | 2193 | 7134530 | |
FQ210155.1 | - | 14, 15 | 2259 |
