siRNA Details
Gene Symbol: Retnlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181625.1 | NP_853656.1 | 1, 2 | 132 | ||
XM_006248282.2 | XP_006248344.1 | 2, 3 | 225 | |||
GenBank mRNA | AJ514932.1 | - | 1, 2 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
