siRNA Details
Gene Symbol: Nit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034126.1 | NP_001029298.1 | 4 | 358 | ||
GenBank mRNA | BC082034.1 | - | 4 | 358 | ||
BC100637.1 | - | 4 | 358 | 5621093 | ||
FQ209436.1 | - | 4 | 366 | |||
FQ219054.1 | - | 1 | 290 | |||
FQ219183.1 | - | 4 | 364 | |||
FQ229934.1 | - | 4 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
