siRNA Details
Gene Symbol: Tmem30c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105889.1 | NP_001099359.1 | 5, 6 | 594 | ||
XM_008768632.1 | XP_008766854.1 | 5, 6 | 831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
