siRNA Details
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002802.1 | NP_001002802.1 | 3 | 508 | ||
XM_006248153.2 | XP_006248215.1 | 3 | 562 | |||
GenBank mRNA | BC166597.1 | - | 3 | 522 | 7936295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
