siRNA Details
Gene Symbol: Hlcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006248111.2 | XP_006248173.1 | 10 | 3127 | ||
XM_006248112.2 | XP_006248174.1 | 10 | 3194 | |||
XM_006248114.2 | XP_006248176.1 | 9 | 2988 | |||
XM_008768611.1 | XP_008766833.1 | 10 | 2326 | |||
XM_008768612.1 | XP_008766834.1 | 9 | 2191 | |||
XM_221630.8 | XP_221630.6 | 9 | 2307 |
