siRNA Details
Gene Symbol: Chaf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024741.1 | NP_001019912.1 | 12 | 1253 | ||
XM_006248044.2 | XP_006248106.1 | 12 | 1289 | |||
GenBank mRNA | BC097460.1 | - | 12 | 1253 | 7457454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
