siRNA Details
Gene Symbol: Mis18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127523.2 | NP_001120995.2 | 4 | 571 | ||
GenBank mRNA | BC167102.1 | - | 4 | 571 | 8373935 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
