siRNA Details
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105897.2 | NP_001099367.2 | 11 | 1290 | ||
GenBank mRNA | CO400743.1 | - | 5 | 483 | ||
FQ219784.1 | - | 3 | 177 | |||
FQ225223.1 | - | 11 | 1263 | |||
FQ226011.1 | - | 11 | 1263 |
