siRNA Details
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105897.2 | NP_001099367.2 | 5 | 629 | ||
GenBank mRNA | CV076369.1 | - | 2 | 207 | ||
DY312304.1 | - | 5 | 629 | |||
FQ225223.1 | - | 5 | 602 | |||
FQ226011.1 | - | 5 | 602 |
