siRNA Details
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100501.1 | NP_001093971.1 | 7 | 826 | ||
XM_006248015.2 | XP_006248077.1 | 7 | 842 | |||
GenBank mRNA | BC087705.1 | - | 7 | 769 | ||
BC112386.1 | - | 7 | 776 | |||
FQ221582.1 | - | 7 | 781 |
