siRNA Details
Gene Symbol: Hsph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011901.1 | NP_001011901.1 | 10, 11 | 1581 | ||
XM_008768947.1 | XP_008767169.1 | 9, 10 | 1342 | |||
GenBank mRNA | BC081945.1 | - | 10, 11 | 1581 | 7121434 |
