siRNA Details
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105906.2 | NP_001099376.1 | 5 | 1127 | ||
NM_001198555.1 | NP_001185484.1 | 5 | 1127 | |||
XM_006248810.2 | XP_006248872.1 | 4 | 1070 | |||
GenBank mRNA | CO396482.1 | - | 1 | 40 | ||
DY309833.1 | - | 2 | 548 |
