siRNA Details
Gene Symbol: LOC288521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013868.1 | NP_001013890.1 | 1339 | |||
GenBank mRNA | BC079301.1 | - | 1339 | 7134538 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC288521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013868.1 | NP_001013890.1 | 1339 | |||
GenBank mRNA | BC079301.1 | - | 1339 | 7134538 |