siRNA Details
Gene Symbol: Akap17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127246.1 | NP_001120718.1 | 5 | 1965 | ||
XM_006248998.2 | XP_006249060.1 | 5 | 2678 | |||
XM_006248999.2 | XP_006249061.1 | 5 | 2522 | |||
XM_006249000.2 | XP_006249062.1 | 5 | 2613 | |||
GenBank mRNA | BC166722.1 | - | 5 | 1965 | 7933458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
