siRNA Details
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004203.3 | NP_001004203.3 | 8 | 1093 | ||
XM_006249005.2 | XP_006249067.1 | 8 | 1215 | |||
XM_006249006.2 | XP_006249068.1 | 8 | 1098 | |||
GenBank mRNA | BC078973.1 | - | 8 | 1093 | 7111847 | |
DQ278199.1 | - | 8 | 1141 | |||
DQ278200.1 | - | 1686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
