siRNA Details
Gene Symbol: RGD1305455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024969.2 | NP_001020140.2 | 5 | 922 | ||
GenBank mRNA | BC091263.1 | - | 4 | 792 | ||
DY317491.1 | - | 290 | ||||
FQ211718.1 | - | 5 | 968 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_153161, BC037034, AK046882, AK138960, CD354602.
