siRNA Details
Gene Symbol: Mospd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025629.1 | NP_001020800.1 | 2 | 362 | ||
XM_006249115.2 | XP_006249177.1 | 3 | 422 | |||
GenBank mRNA | BC099230.1 | - | 2 | 362 | 7442803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
