siRNA Details
Gene Symbol: Actl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105917.1 | NP_001099387.2 | 12 | 1161 | ||
XM_006249120.2 | XP_006249182.1 | 12 | 1242 |

More Information
Other Species Targeted

Gene Ontology Categories:
