siRNA Details
Gene Symbol: Mrps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105923.1 | NP_001099393.1 | 2, 3 | 488 | ||
XM_006249264.2 | XP_006249326.1 | 2, 3 | 208 | |||
GenBank mRNA | FQ223326.1 | - | 2, 3 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
