siRNA Details
Gene Symbol: Arl6ip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025630.1 | NP_001020801.1 | 4 | 932 | ||
GenBank mRNA | BC097374.1 | - | 4 | 932 | 7444619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
