siRNA Details
Gene Symbol: Vps37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105928.1 | NP_001099398.1 | 2 | 303 | ||
XM_006249332.2 | XP_006249394.1 | 1 | 786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
