siRNA Details
Gene Symbol: Cux2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271380.1 | NP_001258309.1 | 17 | 2178 | ||
XM_008769215.1 | XP_008767437.1 | 12 | 1823 | |||
XM_008769216.1 | XP_008767438.1 | 12 | 1802 | |||
XM_008769217.1 | XP_008767439.1 | 12 | 1790 | |||
XM_008769218.1 | XP_008767440.1 | 12 | 1769 | |||
XM_008769219.1 | XP_008767441.1 | 16 | 3279 |
