siRNA Details
Gene Symbol: Cux2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271380.1 | NP_001258309.1 | 5 | 532 | ||
XM_008769219.1 | XP_008767441.1 | 4 | 1633 |

More Information
Other Species Targeted

Gene Ontology Categories:
