siRNA Details
Gene Symbol: Vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105932.1 | NP_001099402.1 | 2, 3 | 286 | ||
XM_006249362.2 | XP_006249424.1 | 3, 4 | 312 | |||
GenBank mRNA | AB578912.1 | - | 1, 2 | 36 | ||
BC167055.1 | - | 3, 4 | 273 | 7302374 | ||
FQ224287.1 | - | 3, 4 | 294 |
