siRNA Details
Gene Symbol: Fam216a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008290.1 | NP_001008291.1 | 7 | 929 | ||
GenBank mRNA | BC086384.1 | - | 7 | 929 | 7301624 | |
FQ220218.1 | - | 7 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
