siRNA Details
Gene Symbol: Rita1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044226.1 | NP_001037691.1 | 3 | 579 | ||
XM_006249383.2 | XP_006249445.1 | 3 | 682 | |||
XM_006249384.2 | XP_006249446.1 | 2 | 557 | |||
GenBank mRNA | BC112384.1 | - | 3 | 579 | 7932995 |

More Information
Other Species Targeted

Gene Ontology Categories:
