siRNA Details
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013046.1 | NP_001013064.1 | 2 | 177 | ||
XM_008769221.1 | XP_008767443.1 | 2 | 227 | |||
GenBank mRNA | BC085769.1 | - | 2 | 177 | 7135343 |

More Information
Other Species Targeted

Gene Ontology Categories:
