siRNA Details
Gene Symbol: RGD1311899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009630.1 | NP_001009630.1 | 3, 4 | 303 | ||
XM_006249433.2 | XP_006249495.1 | 3, 4 | 334 | |||
GenBank mRNA | BC088751.1 | - | 3, 4 | 303 | 7317722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
