siRNA Details
Gene Symbol: Foxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105935.1 | NP_001099405.1 | 10, 11 | 1253 | ||
XM_006249493.2 | XP_006249555.1 | 10, 11 | 1794 |

More Information
Other Species Targeted

Gene Ontology Categories:
