siRNA Details
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013047.1 | NP_001013065.1 | 7 | 558 | ||
XM_008764483.1 | XP_008762705.1 | 6 | 591 | |||
XM_008764484.1 | XP_008762706.1 | 6 | 591 | |||
GenBank mRNA | BC079400.1 | - | 7 | 705 | ||
BC091225.1 | - | 7 | 558 | 7129875 | ||
FQ224358.1 | - | 6 | 596 |
