siRNA Details
Gene Symbol: Spryd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037765.1 | NP_001032854.1 | 1, 2 | 94 | ||
XM_006240746.1 | XP_006240808.1 | 2, 3 | 270 | |||
GenBank mRNA | BC099138.1 | - | 1, 2 | 94 | 7456187 |

More Information
Other Species Targeted

Gene Ontology Categories:
