siRNA Details
Gene Symbol: Stat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011905.1 | NP_001011905.1 | 3 | 367 | ||
XM_006240747.2 | XP_006240809.1 | 3 | 663 | |||
XM_006240748.2 | XP_006240810.1 | 3 | 663 | |||
GenBank mRNA | BC083867.1 | - | 3 | 367 | 7127907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
